4-AZIDO-4-DEOXY-D-GLUCOSE 100MG
Catalog # 77888-728
Supplier: AFG Bioscience
New Product
4-AZIDO-4-DEOXY-D-GLUCOSE 100MG
Catalog # 77888-728
Supplier: AFG Bioscience
Supplier Number: C51534-100MG
New Product
Frequently Bought Together
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
SYRINGE FOR INSULIN WITH NEEDLE 1ML
Supplier: AMX INDUSTRIAL TRADE MX
Bacto™ Brain Heart Infusion, BD Diagnostics
Supplier: BD
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Supplier: TCI America
Orion™ ISE Calibration Standards, Thermo Scientific
Supplier: Thermo Fisher Scientific
Sol-Guard® Safety TB Syringes, Sol M
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Pierce Zeba™ Desalt Spin Columns, Thermo Scientific
Supplier: Thermo Scientific
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Calcium chloride, dried 97%, powder
Supplier: Thermo Scientific Chemicals
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Endothelial Cell Growth Media Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA