234-TRI-O-BENZOYL-Α-D-GLCRN 250MG
Catalog # 77888-963
Supplier: AFG Bioscience
New Product
234-TRI-O-BENZOYL-Α-D-GLCRN 250MG
Catalog # 77888-963
Supplier: AFG Bioscience
Supplier Number: C53981-250MG
New Product
Frequently Bought Together
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Supplier: Thermo Scientific
Sodium chloride ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
VWR® Razor Blades
Supplier: VWR International
Betaine enhancer solution 5M
Supplier: VWR International
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Reagent alcohol
Supplier: VWR International
Pierce™ Blocker™ Casein in PBS, Thermo Scientific
Supplier: Thermo Scientific
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
CL17 DPD Indicator, Hach
Supplier: Hach
Sodium phenoxide 98%
Supplier: Thermo Scientific Chemicals
Sodium peroxodisulfate 98%
Supplier: Thermo Scientific Chemicals
Chloroform ≥99.8% stabilised, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Water, deionised
Supplier: Hach