-GAL-PEG3-THIOL 25MG
Catalog # 77888-903
Supplier: AFG Bioscience
New Product
-GAL-PEG3-THIOL 25MG
Catalog # 77888-903
Supplier: AFG Bioscience
Supplier Number: C53013-25MG
New Product
Frequently Bought Together
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
Leakbuster™ Specimen Containers
Supplier: Starplex
Oakton® Conductivity and TDS Standards, Cole Parmer
Supplier: Antyila Scientific
Methanol, anhydrous ≥99.8% (by GC), GR ACS ACS, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Preserved Starfish
Supplier: Avantor
SYRINGE FOR INSULIN WITH NEEDLE 1ML
Supplier: AMX INDUSTRIAL TRADE MX
Guanidinium hydrochloride, Technical Grade
Supplier: VWR
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Sol-Guard® Safety TB Syringes, Sol M
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8-100.1% (dried basis) ACS
Supplier: Thermo Scientific Chemicals
CIP 100® Alkaline Process and Research Cleaners
Supplier: Steris
LabKlenz™ 250 Acid Detergents
Supplier: Steris
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Tri-Corn Beakers, Plastic, Disposable, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Glycerine 99+%
Supplier: Thermo Scientific Chemicals
Ward's® Live <i>Lactobacillus delbrueckii ss. bulgaricus</i> Culture
Supplier: Avantor