BUFFER PYROSOL W/PH 5.5ML
Catalog # MSPP-BR05125
Supplier: ASSOCIATES OF CAPE COD INC MS
BUFFER PYROSOL W/PH 5.5ML
Catalog # MSPP-BR05125
Supplier: ASSOCIATES OF CAPE COD INC MS
Supplier Number: BR05125
Frequently Bought Together
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
Calcium chloride dihydrate 99.0-107.0% USP, Ph. Eur., BP, JP endotoxin tested
Supplier: Avantor Performance Materials
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Glycerine ≥99.7% ACS, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
qScript™ XLT One-Step RT-PCR Kit, QuantaBio
Supplier: Quantabio
Sandwich Bags, Low Density Flip Top Elkay Plastics
Supplier: Elkay Plastics
qScript™ cDNA SuperMix, QuantaBio
Supplier: Quantabio
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics