Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Recombinant TGF-β1 PLUS (from E. coli)

Human Recombinant TGF-β1 PLUS (from E. coli)

Vial, 0.025 mg
Catalog # 77870-773
Supplier:   Sartorius
Supplier #:  CYK-0025-0010
Human Recombinant TGF-β1 PLUS (from E. coli)
Catalog # 77870-773
Supplier:  Sartorius
CAS Number:  
Human Recombinant TGF-β1 PLUS (from E. coli)
Human Recombinant TGF-β1 PLUS (from E. coli)

Vial, 0.025 mg

CAS Number:  
Price
$668.34
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.

Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    0.025 mg
  • Protease-free:
    N
  • Protein/peptide name:
    TGF-β1 PLUS
  • Protein synonyms:
    Transforming Growth Factor-β1 PLUS
  • Protein Function:
    Cytokine
  • Source:
    E. coli
  • Shipping temperature:
    Ambient
  • Nuclease-free:
    N
  • Cat. No.:
    77870-773
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Ambient
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Endotoxin-free:
    Y
  • Species:
    Human
  • Animal-free:
    Y

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.