Human Recombinant BDNF (from E. coli)
Vial, 0.1 mg
Catalog # 77870-823
Supplier: Sartorius
Supplier #: CYK-0100-0008
Jump to:
Product Details & Documents
Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.
Documents
TDS RUO Growth Factors Cytokines.pdf
Datasheet
Specifications
Specifications
- Pack type:Vial
- Size:0.1 mg
- Protease-free:N
- Protein/peptide name:BDNF
- Protein synonyms:Brain Derived Neurotrophic Factor
- Protein Function:Cytokine
- Source:E. coli
- Shipping temperature:Ambient
- Nuclease-free:N
- Cat. No.:77870-823
- Protein/peptide type:Recombinant
- Storage conditions:Ambient
- Carrier-free:Y
- Conjugation:Unconjugated
- Endotoxin-free:Y
- Species:Human
- Animal-free:Y



