Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Recombinant BDNF (from E. coli)

Human Recombinant BDNF (from E. coli)

Vial, 0.5 mg
Catalog # 77870-871
Supplier:   Sartorius
Supplier #:  CYK-0500-0008
Human Recombinant BDNF (from E. coli)
Catalog # 77870-871
Supplier:  Sartorius
CAS Number:  
Human Recombinant BDNF (from E. coli)
Human Recombinant BDNF (from E. coli)

Vial, 0.5 mg

CAS Number:  
Price
$3,760.40
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.

Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    0.5 mg
  • Protease-free:
    N
  • Protein/peptide name:
    BDNF
  • Protein synonyms:
    Brain Derived Neurotrophic Factor
  • Protein Function:
    Cytokine
  • Source:
    E. coli
  • Shipping temperature:
    Ambient
  • Nuclease-free:
    N
  • Cat. No.:
    77870-871
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Ambient
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Endotoxin-free:
    Y
  • Species:
    Human
  • Animal-free:
    Y

Specifications