HAIR BEARD NETS EAR PLUGS ZWB L CS43
Catalog # 7786767-924
Supplier: Terracycle
New Product
HAIR BEARD NETS EAR PLUGS ZWB L CS43
Catalog # 7786767-924
Supplier: Terracycle
Supplier Number: CS43-L
New Product
Frequently Bought Together
PolarPack® Water Based Foam Bricks, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
VWR® Sharps Containers with Flip Up Lids
Supplier: VWR International
Dual-Tested Autoclave Biohazardous Waste Bags, Medegen Medical Products
Supplier: Medegen Medical Products, LLC
Infecon® Absorbent Strips, Com-Pac
Supplier: Com-Pac
Lab Mailers and Mailing Sleeves for Specimen Transport, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
VWR® Sharps Container Systems, Multiple Colors and Styles
Supplier: VWR International
VWR® Sharps Containers with Sliding Lid
Supplier: VWR International
Insulated Shippers, Expanded Polystyrene (EPS), Foam Only and with Corrugate
Supplier: Sonoco Thermosafe
PAPER PRINTER ROLL PK3
Supplier: STERIS CORP MS
Sterilization Pouches, Heat Seal Paper
Supplier: O&M HALYARD CANADA, INC CA
Shipper, Category B Refrigerated, Medium 95 kPa Canister, Therapak®
Supplier: Therapak, LLC
Beaker with Dual Temp Strip, 6.5 oz., Therapak
Supplier: Therapak, LLC
SHARPS COLLECTR 7.6L FUNL CS24
Supplier: Avantor
Biohazardous Waste Bags, Medegen Medical Products
Supplier: Medegen Medical Products, LLC