ANTIBODY RB X-LMTK2 TRIAL A304-270A-T
Catalog # 77788-535
Supplier: BETHYL LABORATORIES
New Product
ANTIBODY RB X-LMTK2 TRIAL A304-270A-T
Catalog # 77788-535
Supplier: BETHYL LABORATORIES
Supplier Number: A304-270A-T
New Product
Frequently Bought Together
SYRINGE FOR INSULIN WITH NEEDLE 1ML
Supplier: AMX INDUSTRIAL TRADE MX
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
1X PBS (Phosphate Buffered Saline), pH 7.4
Supplier: GROWCELLS
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Hydrochloric acid 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Supplier: TCI America
Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: Genetex
Potassium Sulfate
Supplier: Ward's Science
DAPT ≥98% (by NMR)
Supplier: Enzo Life Sciences
Difco™ Baird-Parker Agar Base, BD
Supplier: BD
tri-Sodium citrate dihydrate ≥99% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Bovine Serum Albumin (BSA)
Supplier: MilliporeSigma
Human Recombinant CNTF (from <i>E. coli</i>)
Supplier: Peprotech
Sol-M® Standard Syringes without Needle
Supplier: SOL-MILLENNIUM CANADA, INC. CA