ANTIBODY RB X-LMTK2 A304-270A
Catalog # 77788-534
Supplier: BETHYL LABORATORIES
New Product
ANTIBODY RB X-LMTK2 A304-270A
Catalog # 77788-534
Supplier: BETHYL LABORATORIES
Supplier Number: A304-270A
New Product
Frequently Bought Together
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
Phenolphthalein 1% (w/v) in 95% (v/v) alcohol indicator, neutralized
Supplier: Ricca Chemical
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Extracta™ DNA Prep for PCR, Quantabio
Supplier: Quantabio
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Supplier: MilliporeSigma
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Quality Economy Brand Luer Slip Syringes, Air-Tite Products
Supplier: Air-Tite
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
BPMO ≥99% (by 1H-NMR), high purity
Supplier: Enzo Life Sciences
Pap-Pen; Slide Marker, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
tri-Sodium citrate dihydrate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Supplier: TCI America
Sol-Vet® Blood Collection Needles (Veterinary Care), Sol M
Supplier: SOL-MILLENNIUM CANADA, INC. CA
BBL™ SFP Agar Base, BD
Supplier: BD
Silicone oil for oil baths –40…+200°C
Supplier: Thermo Scientific Chemicals