ANTIBODY RB X-FOXC1 TRIAL A303-520A-T
Catalog # 77787-157
Supplier: BETHYL LABORATORIES
New Product
ANTIBODY RB X-FOXC1 TRIAL A303-520A-T
Catalog # 77787-157
Supplier: BETHYL LABORATORIES
Supplier Number: A303-520A-T
New Product
Frequently Bought Together
Buffers, Precision Reference Standards, RICCA Chemical Company
Supplier: Ricca Chemical
Sol-M® Standard Syringes without Needle
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Preserved Starfish
Supplier: Avantor
Eveready® Alkaline Batteries, Bulbtronics
Supplier: Bulbtronics
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Molecular sieve 3A (0.3 nm, 3 Å), beads 1-2 mm (0.04-0.08 in)
Supplier: Thermo Scientific Chemicals
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Acetyl-L-carnitine hydrochloride
Supplier: Thermo Scientific Chemicals
Magnesium chloride, anhydrous 99%
Supplier: Thermo Scientific Chemicals
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
Bromothymol blue 0.04% (w/v) in aqueous solution indicator
Supplier: Ricca Chemical
Sera-Xtracta Virus/Pathogen Kits, Cytiva
Supplier: Cytiva
DNA MINICOLUMNS V-SPIN HIBIND PK1000
Supplier: Omega Bio-Tek
Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International
Sandwich Bags, Low Density Flip Top Elkay Plastics
Supplier: Elkay Plastics