ANTIBODY RB X-FOXC1 A303-520A
Catalog # 77787-156
Supplier: BETHYL LABORATORIES
New Product
ANTIBODY RB X-FOXC1 A303-520A
Catalog # 77787-156
Supplier: BETHYL LABORATORIES
Supplier Number: A303-520A
New Product
Frequently Bought Together
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Medicine Cups, DUKAL™ Corporation
Supplier: Dukal
Bacto™ Brain Heart Infusion, BD Diagnostics
Supplier: BD
Polyethylene Disposable Apron
Supplier: Dival Safety & Supplies
Leakbuster™ Specimen Containers
Supplier: Starplex
Bulk Syringes, Nonsterile, BD Medical
Supplier: BD
Coagulase
Supplier: BD
Ward's® Live <i>Escherichia coli</i> JM101 (Host) Culture
Supplier: Avantor
pH Buffer Capsules, Oakton® Instruments
Supplier: Antyila Scientific
Diisopropyl azodicarboxylate 94%
Supplier: Thermo Scientific Chemicals
Hydrazinium sulfate ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Supplier: TCI America
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Dimethyl sulfoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®
Supplier: VWR International
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
tri-Sodium citrate dihydrate ≥99% (by non aqueous titration), granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC