ANTIBODY SPKYG 593 ANTI-HU CD8 25TEST
Catalog # CA77782-204
Supplier: Biolegend
New Product
ANTIBODY SPKYG 593 ANTI-HU CD8 25TEST
Catalog # CA77782-204
Supplier: Biolegend
Supplier Number: 344789
New Product
Frequently Bought Together
BD Angiocath™ IV Catheters, BD™
Supplier: BD
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
COD Digestion vials, VWR Chemicals BDH®
Supplier: VWR International
Preserved Starfish
Supplier: Avantor
DIBMA Screening Kit, HEPES
Supplier: CUBE BIOTECH
Kerosene, odorless
Supplier: INNOVA CHEMICALS CA
Specialty Cell Culture Media
Supplier: Lonza
Brucella Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
4,4'-Isopropylidenediphenol 97+%
Supplier: Thermo Scientific Chemicals
E.Z.N.A.® Plasmid Mini Kit I, Omega Bio-tek®
Supplier: Omega Bio-Tek
Sodium Phosphate Dibasic Anhydrous
Supplier: Ward's Science
Ricinoleic acid ≥80.0%
Supplier: TCI America
Hydroxylammonium chloride ACS, ISO, Supelco®
Supplier: MilliporeSigma
Acetic acid glacial 99.5-100.5% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Ponceau S solution 0.1% in 1% acetic acid protein stain
Supplier: Azure Biosystems
Cutting Mats, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
1,8-Diazabicyclo[5.4.0]undec-7-ene for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
ZymoTaq DNA Polymerase, Zymo Research
Supplier: Zymo Research
Hardness 1 Buffer Solution, Hach
Supplier: Hach
pH Buffer Capsules, Oakton® Instruments
Supplier: Antyila Scientific