ANTIBODY GMP ULEAF ANTI-HU CD34 100UG.
CA77782-107
New Product
Frequently Bought Together
Acetonitrile ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
: MilliporeSigma
VWR® Razor Blades
: VWR International
Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
: VWR International
B352 Specimen Bottles and Caps, Simport Scientific
: Simport Scientific
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
Ethanol denatured, anhydrous
: AVANTOR PERFORMANCE MATERIAL LLC
Corning® RPMI 1640, Corning
: Corning
Sodium molybdate(VI) dihydrate 99.5-103.0% ACS
: Thermo Scientific Chemicals
High Sensitivity RNA ScreenTape Analysis
: AGILENT TECHNOLOGIES (GENOMICS) CA
MycoAlert™ Mycoplasma Detection Kits, Lonza
: LONZA PHARMA - BIOSCIENCE CA