ANTIBODY ULEAF ANTI-HU CD122 IL-2RΒ 1MG
Catalog # CA77782-005
Supplier: Biolegend
New Product
ANTIBODY ULEAF ANTI-HU CD122 IL-2RΒ 1MG
Catalog # CA77782-005
Supplier: Biolegend
Supplier Number: 339024
New Product
Frequently Bought Together
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
TUBE K2EDTA MINICOLLECT 0.25/0.5ML PK500
Supplier: Greiner Bio-One
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
THREADLOCKER 222 BOTTLE 50ML
Supplier: Loctite
VWR® Reagent alcohol
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
n-Heptane ≥99% (n-heptane) [≥99.9% (saturated C7 hydrocarbons)], OmniSolv®, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Fluoride Standards, 1 ml = 1 mg F-, 1000 ppm F-, RICCA Chemical Company
Supplier: Ricca Chemical
Fmoc-6-Ahx-OH 98%
Supplier: Thermo Scientific Chemicals
VWR® Unisex Staff Lab Coat
Supplier: VWR International
Biebrich scarlet
Supplier: Thermo Scientific Chemicals
Costar® Brand Cell Culture Clusters, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Pharmacy Bags, Reclosable, Elkay Plastics
Supplier: Elkay Plastics
N,N-Dimethylformamide ≥99.8% (by GC) for peptide synthesis, Burdick & Jackson™
Supplier: Burdick & Jackson