ANTIBODY 520 ANHU CD137 4-1BB 100TEST
Catalog # CA77780-962
Supplier: Biolegend
New Product
ANTIBODY 520 ANHU CD137 4-1BB 100TEST
Catalog # CA77780-962
Supplier: Biolegend
Supplier Number: 300816
New Product
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Leakbuster™ Specimen Containers
Supplier: Starplex
VWR® Specimen Containers, PP with PE Caps
Supplier: VWR International
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
Calcium hydroxide, Technical Grade
Supplier: VWR
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
THREADLOCKER 222 BOTTLE 50ML
Supplier: Loctite
HardyVal™ Tryptic Soy Broth, USP, Media-Bag™, Hardy Diagnostics
Supplier: Hardy Diagnostics
E. cloni® Chemically Competent Cells, Biosearch Technologies
Supplier: Lucigen
HISTOSETTE® II Cassettes in QuickLoad ™ Sleeve, Simport
Supplier: Simport Scientific
BPMO ≥99% (by 1H-NMR), high purity
Supplier: Enzo Life Sciences
Molecular sieve 3A (0.3 nm, 3 Å), pellets Mesh 8 - 12, Ø 1/16", Supelco®
Supplier: MilliporeSigma
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA