MONOMER FXT-B SRCV2 LLYDANYFL 25UG
Catalog # CA77780-897
Supplier: Biolegend
New Product
MONOMER FXT-B SRCV2 LLYDANYFL 25UG
Catalog # CA77780-897
Supplier: Biolegend
Supplier Number: 280079
New Product
Frequently Bought Together
Hydrion™, Mikro pH Test Paper with Dispensor, Micro Essential Laboratory
Supplier: Micro Essential
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Razor Blades
Supplier: VWR International
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
Acetone ACS for production, VWR Chemicals BDH®
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: Chemetrics
Tergazyme® Enzyme-Active Powder Detergents
Supplier: Alconox
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
Toluene ACS
Supplier: INNOVA CHEMICALS CA