GMP PE/CYANINE7 ANTI-HU CD7 100TEST
Catalog # CA77780-813
Supplier: Biolegend
New Product
GMP PE/CYANINE7 ANTI-HU CD7 100TEST
Catalog # CA77780-813
Supplier: Biolegend
Supplier Number: 260296
New Product
Frequently Bought Together
Accessories for Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Buffer Solutions
Supplier: VWR
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Specialty Cell Culture Media
Supplier: Lonza
Molecular sieve 3A (0.3 nm, 3 Å), pellets Mesh 8 - 12, Ø 1/16", Supelco®
Supplier: MilliporeSigma
Water USP, Ph. Eur., purified
Supplier: Ricca Chemical
Thioacetamide ≥99% ACS
Supplier: Thermo Scientific Chemicals
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
Hexachloroplatinic(IV) acid hexahydrate Pt ∼ 37,5%, BAKER ANALYZED® ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Sodium Bicarbonate
Supplier: Ward's Science