ANTIBODY GMP PE ANTI-HU CD14 100TEST
Catalog # CA77780-744
Supplier: Biolegend
New Product
ANTIBODY GMP PE ANTI-HU CD14 100TEST
Catalog # CA77780-744
Supplier: Biolegend
Supplier Number: 260112
New Product
Frequently Bought Together
Masterflex® Transfer Tubing, C-Flex®, Clear, Avantor®
Supplier: VWR International
Sodium hydroxide, pellets ACS
Supplier: VWR
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Accessories for Osmometer, Advanced Instruments
Supplier: Advanced Instruments
Petroleum spirit, 35…60 °C, GR ACS, Supelco®
Supplier: MilliporeSigma
Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
TEGADERM DRESSING 6X7CM 100'S
Supplier: Safecross First Aid Limited
Optical Lens Cleaner, FoxWest Sales
Supplier: Decon Labs
VWR® Proteinase K
Supplier: VWR
Sodium thiosulfate 0.100 N (N/10) in aqueous solution
Supplier: Ricca Chemical
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
TelN Protelomerase
Supplier: New England Biolabs (NEB)
Cryoscope Standards, Advanced Instruments
Supplier: Advanced Instruments
Difco™ YPD Medium, BD Life Sciences
Supplier: BD
Phosphate Buffered Saline
Supplier: Ward's Science
Sandwich Bags, Low Density Flip Top Elkay Plastics
Supplier: Elkay Plastics