ANTIBODY GMP PERCP ANTI-HU CD45 100TEST
Catalog # CA77780-743
Supplier: Biolegend
New Product
ANTIBODY GMP PERCP ANTI-HU CD45 100TEST
Catalog # CA77780-743
Supplier: Biolegend
Supplier Number: 260110
New Product
Frequently Bought Together
Starswab® Anaerobic Transport System, Starplex®
Supplier: Starplex
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Silver nitrate 0.100 N (N/10) in aqueous solution
Supplier: Ricca Chemical
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: Chemetrics
Puritan® MALDI-TOF Pointed Applicator Stick, Puritan Medical Products
Supplier: Puritan Medical Products
ES 7X™ Concentrate, MP Biomedicals
Supplier: MP Biomedicals
Enzymes and Separation Accessories, Omega Bio-Tek
Supplier: Omega Bio-Tek
STORAGE SOLUTION PH ELECTRODE
Supplier: Oakton
Conflikt® Ready-to-use Disinfectant
Supplier: Decon Labs