ANTIBODY GMP PB ANTI-HU CD45 100TEST
Catalog # CA77780-742
Supplier: Biolegend
New Product
ANTIBODY GMP PB ANTI-HU CD45 100TEST
Catalog # CA77780-742
Supplier: Biolegend
Supplier Number: 260108
New Product
Frequently Bought Together
Caliber™ Type VII Gauze Sponges, DUKAL™ Corporation
Supplier: Dukal
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Bacto™ Brain Heart Infusion, BD Diagnostics
Supplier: BD
Potassium chloride saturated aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
Leakbuster™ Specimen Containers
Supplier: Starplex
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
Orthophosphoric acid ≥85%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
n-Hexane ≥95%, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Hydrazinium sulfate ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
AGAR BRAIN HEART INFUSION 450 G
Supplier: BECTON DICKINSON DE MEXICO, S. MX
EMS Swiss Line Standard Tweezer Style 2A, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
EE Broth Mossel
Supplier: HARDY DIAGNOSTICS CA
VACUUM PUMP VACUUBRAND ME1C
Supplier: Brandtech
Manganese(II) Chloride
Supplier: Ward's Science
Human Recombinant FGF-basic (from E. coli)
Supplier: Peprotech
ACK Lysing Buffer, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.