ANTIBODY A1272 ANHUM CD129 (IL-9 R) 10UG
Catalog # CA77781-380
Supplier: Biolegend
New Product
ANTIBODY A1272 ANHUM CD129 (IL-9 R) 10UG
Catalog # CA77781-380
Supplier: Biolegend
Supplier Number: 310413
New Product
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Sodium chloride ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
VWR® Specimen Containers with Dual Click
Supplier: VWR International
Water for HPLC, Supelco®
Supplier: MilliporeSigma
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
pH Buffer Capsules, Oakton® Instruments
Supplier: Antyila Scientific
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Supplier: TCI America
Axygen® Graduated Transport Tubes, Corning
Supplier: Corning
Total Nitrogen Reagent Sets, HR, TNT
Supplier: Hach
VWR® Gas Pressure Regulators
Supplier: VWR International
Pharmacy Bags, Reclosable, Elkay Plastics
Supplier: Elkay Plastics
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Acetic acid glacial 99.5-100.5% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Hazmat Boot Covers, Onguard
Supplier: Onguard Industries