ANTIBODY GMP PE ANTI-HU CD20 100TEST
Catalog # CA77780-839
Supplier: Biolegend
New Product
ANTIBODY GMP PE ANTI-HU CD20 100TEST
Catalog # CA77780-839
Supplier: Biolegend
Supplier Number: 260352
New Product
Frequently Bought Together
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Supplier: Ricca Chemical
SOLUTION ELER CLNG 450 ML INORGANIC SUBS
Supplier: Hanna
Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Supplier: Starplex
Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Reagent alcohol
Supplier: VWR International
Fendall® Saline Eyewash Bottles, Honeywell Safety
Supplier: Honeywell Safety Products
Sodium hydroxide 0.100 N (N/10) in aqueous solution
Supplier: Ricca Chemical
Nitric acid 65%, Suprapur® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
Circular Cover Glass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Sodium Phosphate Dibasic Anhydrous
Supplier: Ward's Science
Hazmat Boot Covers, Onguard
Supplier: Onguard Industries
Pierce™ Blocker™ Casein in PBS, Thermo Scientific
Supplier: Thermo Scientific
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International
HYDRAVER 2 HYDRAZINE RG 1000ML
Supplier: Hach