GMP APC ANTI-HU CD117 (C-KIT) 100TEST
Catalog # CA77780-748
Supplier: Biolegend
New Product
GMP APC ANTI-HU CD117 (C-KIT) 100TEST
Catalog # CA77780-748
Supplier: Biolegend
Supplier Number: 260120
New Product
Frequently Bought Together
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
Hydrochloric acid 36.5 - 38.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
Bovine Serum Albumin (BSA)
Supplier: VWR
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Quality Economy Brand Luer Slip Syringes, Air-Tite
Supplier: Air-Tite
Quality Economy Brand Luer Lock Syringes, Air-Tite
Supplier: Air-Tite
Acetonitrile ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
InLab® Intelligent Sensor Management (ISM®) pH/Conductivity Electrodes, METTLER TOLEDO
Supplier: Mettler Toledo
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
Optical Lens Cleaner, FoxWest Sales
Supplier: Decon Labs
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
HistoGel™ Specimen Processing Gel, Epredia
Supplier: Thermo Scientific
Orion™ Maintenance Solutions for Sodium Electrodes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Succinimide 98+%
Supplier: Thermo Scientific Chemicals
Tissue Marking Dyes (TMD™), TBS®, General Data
Supplier: General Data Healthcare
Difco™ Baird-Parker Agar Base, BD
Supplier: BD