ANTIBODY GMP ANTI-HU CD38 100TEST
Catalog # CA77780-832
Supplier: Biolegend
New Product
ANTIBODY GMP ANTI-HU CD38 100TEST
Catalog # CA77780-832
Supplier: Biolegend
Supplier Number: 260336
New Product
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Quality Economy Brand Luer Slip Syringes, Air-Tite
Supplier: Air-Tite
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® Razor Blades
Supplier: VWR International
Acetonitrile ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Lecithin, refined
Supplier: Thermo Scientific Chemicals
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
Microsep Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
LabKlenz™ 100 Alkaline Detergents
Supplier: Steris
DNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Supplier: TCI America
DAPT ≥98% (by NMR)
Supplier: Enzo Life Sciences
Sandwich Bags, Low Density Flip Top Elkay Plastics
Supplier: Elkay Plastics
X-tourn™ Tourniquets
Supplier: Heathrow Scientific
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA