APC ANTI-MU/HU CD282 (TLR2) RECOM 25UG
Catalog # CA77780-120
Supplier: Biolegend
New Product
APC ANTI-MU/HU CD282 (TLR2) RECOM 25UG
Catalog # CA77780-120
Supplier: Biolegend
Supplier Number: 153005
New Product
Frequently Bought Together
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Bulk Syringes, Nonsterile, BD Medical
Supplier: BD
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
VWR® ClikSeal Specimen Containers
Supplier: VWR International
Lutetium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
MinION™ Mk1B DNA/RNA Sequencing Device, Oxford Nanopore Technologies
Supplier: Oxford Nanopore Technologies
Accessories for Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Toluene ACS
Supplier: INNOVA CHEMICALS CA
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Alcohol reagent 100%
Supplier: Mortech Manufacturing