ANTIBODY C0565 ANTI-MS MERTK (MER) 10UG
Catalog # CA77780-070
Supplier: Biolegend
New Product
ANTIBODY C0565 ANTI-MS MERTK (MER) 10UG
Catalog # CA77780-070
Supplier: Biolegend
Supplier Number: 151513
New Product
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Citranox® Acid Cleaner and Detergents
Supplier: Alconox
BD Angiocath™ IV Catheters, BD™
Supplier: BD
tert-Butyl methyl ether ≥99.5%, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Masterflex® Adapter Fittings, Male Luer to Hose Barb, Straight, Polypropylene, Avantor®
Supplier: VWR International
2-Propanol ≥99.8% (by GC), HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Supplier: VWR International
VeriSpec™ Iron (Fe) Standards for AAS, RICCA Chemical Company
Supplier: Ricca Chemical
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Ward's® Live <i>Lactobacillus delbrueckii ss. bulgaricus</i> Culture
Supplier: Avantor
Rapid Adapter Auxiliary V14
Supplier: Oxford Nanopore Technologies
Dimethyl sulfoxide, sterile filtered
Supplier: Corning
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics
Toluene ACS
Supplier: INNOVA CHEMICALS CA
Triphenylphosphine ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Phthalic anhydride, anhydrous 99.0-100.2% ACS
Supplier: Thermo Scientific Chemicals