ANTIBODY SPKYG 570 ANTI-MU LY-6G 25UG
Catalog # CA77779-560
Supplier: Biolegend
New Product
ANTIBODY SPKYG 570 ANTI-MU LY-6G 25UG
Catalog # CA77779-560
Supplier: Biolegend
Supplier Number: 127681
New Product
Frequently Bought Together
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Accessories for Osmometer, Advanced Instruments
Supplier: Advanced Instruments
VWR® Razor Blades
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
Sodium hydroxide 5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Supplier: Chemetrics
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Glucose Tolerance Drinks, Azer Scientific
Supplier: Azer Scientific
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
BBL™ SFP Agar Base, BD
Supplier: BD
BUFFER SOLN TOT CHLORINE 473ML
Supplier: Hach
Dieffenbach Bulldog Clamp, OR Grade, Sklar
Supplier: Sklar
Keratinocyte Growth Medium 2, PromoCell
Supplier: Promocell
XbaI
Supplier: New England Biolabs (NEB)
Bovine Serum Albumin (BSA)
Supplier: Rockland Immunochemical