ANTIBODY SPKYG 570 ANTI-MU F4/80 100UG
Catalog # CA77779-422
Supplier: Biolegend
New Product
ANTIBODY SPKYG 570 ANTI-MU F4/80 100UG
Catalog # CA77779-422
Supplier: Biolegend
Supplier Number: 123160
New Product
Frequently Bought Together
Formalin 10%, neutral buffered, VWR®
Supplier: VWR International
Glycerine ≥99.7% ACS, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Disposable Biopsy Punches, Integra™ Miltex®
Supplier: Miltex
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Accessories for Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
VWR® Disposable Petri Dishes
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Histoplex™ Histology Containers, Starplex®
Supplier: Starplex
AGAR BRAIN HEART INFUSION 450 G
Supplier: BECTON DICKINSON DE MEXICO, S. MX
SYRINGE WITH NEEDLE 5ML
Supplier: AMX INDUSTRIAL TRADE MX
BBL™ SFP Agar Base, BD
Supplier: BD
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics
CL17 DPD Indicator, Hach
Supplier: Hach
High Sensitivity DNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Reagent Sets for Colorimeters, Hach
Supplier: Hach