ANTIBODY C0227 ANMU CD122 (IL-2RΒ) 10UG
Catalog # CA77778-982
Supplier: Biolegend
New Product
ANTIBODY C0227 ANMU CD122 (IL-2RΒ) 10UG
Catalog # CA77778-982
Supplier: Biolegend
Supplier Number: 105915
New Product
Frequently Bought Together
SYRINGE FOR INSULIN WITH NEEDLE 1ML
Supplier: AMX INDUSTRIAL TRADE MX
Duracell® Silver Oxide Batteries, Bulbtronics
Supplier: Bulbtronics
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
OneStep™ PCR Inhibitor Removal Kits, Zymo Research
Supplier: Zymo Research
Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Supplier: Starplex
Solvent Resistant Pen, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Potassium bromide, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
Corning® rat collagen I (from tail tendon)
Supplier: Corning
Ward's® Live <i>Lactobacillus delbrueckii ss. bulgaricus</i> Culture
Supplier: Avantor
Accessories for Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International
Corning® Cell Recovery Solution, Corning
Supplier: Corning
pH Buffer Capsules, Oakton® Instruments
Supplier: Antyila Scientific
4-Methylpiperidine 98+%
Supplier: Thermo Scientific Chemicals
Difco™ YPD Medium, BD Life Sciences
Supplier: BD