ANTIBODY B0227 ANMU CD122 (IL-2RΒ) 10UG
Catalog # CA77778-981
Supplier: Biolegend
New Product
ANTIBODY B0227 ANMU CD122 (IL-2RΒ) 10UG
Catalog # CA77778-981
Supplier: Biolegend
Supplier Number: 105913
New Product
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Glycerine ≥99.7% ACS, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
VWR® TBE 10X Liquid Concentrate, Ultra Pure Grade
Supplier: VWR
Methanol, anhydrous ≥99.8% (by GC), GR ACS ACS, Supelco®
Supplier: MilliporeSigma
Quality Economy Brand Luer Lock Syringes, Air-Tite
Supplier: Air-Tite
Labels for Cold/Cryogenically Frozen Surfaces, Brady
Supplier: Avantor
Corning® Matrigel® Basement Membrane Matrix, Corning
Supplier: Corning
Ammonia (Nitrogen) Test Strips, Hach
Supplier: Hach
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Sodium hydroxide 98%, pellets
Supplier: Thermo Scientific Chemicals
Transport Vials, Caplugs
Supplier: Caplugs
Norcamphor 98+%
Supplier: Thermo Scientific Chemicals
Water, HyClone™ HyPure, Molecular Biology Grade
Supplier: Cytiva
Toluene ACS
Supplier: INNOVA CHEMICALS CA
Lactose monohydrate for microbiology, Millipore®
Supplier: MilliporeSigma