ANTIBODY SPKYG 570 ANTI-MU CD8A 25UG
CA77778-739
New Product
Frequently Bought Together
Water, OmniSolv® LC-MS for LC-MS, Supelco®
: MilliporeSigma
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
Leakbuster™ Specimen Containers
: Starplex
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
: Thermo Scientific
B352 Specimen Bottles and Caps, Simport Scientific
: Simport Scientific
Sodium thiosulfate 0.100 N (N/10) in aqueous solution
: Ricca Chemical
Ethanol denatured, anhydrous
: AVANTOR PERFORMANCE MATERIAL LLC
2-Propanol ≥99.8% (by GC), HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
: VWR International
DNA MINICOLUMNS V-SPIN HIBIND PK1000
: Omega Bio-Tek
Calcium Nitrate 4-Hydrate
: Ward's Science
High Sensitivity RNA ScreenTape Analysis
: AGILENT TECHNOLOGIES (GENOMICS) CA
ACETIC RINSE SOLUTION 4L
: Ostrem Chemical
Autoclave Wraps, HPK Industries
: HPK Industries