ANTIBODY SPKYG 570 ANTI-MU CD45 25UG
Catalog # CA77778-845
Supplier: Biolegend
New Product
ANTIBODY SPKYG 570 ANTI-MU CD45 25UG
Catalog # CA77778-845
Supplier: Biolegend
Supplier Number: 103171
New Product
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Quality Economy Brand Luer Lock Syringes, Air-Tite Products
Supplier: Air-Tite
VWR® Blood Bank Saline Solutions
Supplier: VWR International
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
Sulfuric acid 95.0-98.0% ACS
Supplier: Anachemia
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Micro Cover Glasses, Square and Rectangular Sizes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
ACETIC RINSE SOLUTION 4L
Supplier: Ostrem Chemical
Anti-CD252 (OX40L) Mouse Monoclonal Antibody (Biotin) [clone: 11C3.1]
Supplier: Biolegend
DNA MINICOLUMNS V-SPIN HIBIND PK1000
Supplier: Omega Bio-Tek
Toluene ACS
Supplier: INNOVA CHEMICALS CA