Multi-Bucket System for Flat Mops
Product Details & Documents
The multi-bucket system is designed for use with Contec’s flat mops, offering flexibility, mobility, and easy customization for cleanroom cleaning. The system features two or three buckets in stainless steel or polypropylene.
Configurable with two or three buckets in stainless steel or polypropylene
- Extendable front frame accommodates an additional bucket for added capacity
- Quiet, locking casters ensure smooth movement and stability
- Utility handle for easy transport of cleaning supplies
- Integrated mop holders with mop brace for secure storage during transport
- Hands-free mop removal tools are available for VertiKlean® MAX™ and QuickTask™ mop heads
The multi-bucket system is a flexible and customizable solution for cleanroom mopping, designed for use with Contec’s flat mops. Available in stainless steel or polypropylene, it can be configured with two or three buckets, with an extendable front frame to accommodate an additional bucket for increased capacity. Quiet, locking casters ensure smooth movement and stability, while a utility handle and an integrated mop holder provide easy transport and secure storage.
A hands-free mop removal tool is available for VertiKlean® MAX™ and QuickTask™ mop heads, enhancing efficiency. Compatible with Contec’s stainless steel downpress wringer for flat mops, the system is autoclavable at 121 °C for 30 minutes, making it ideal for sterilization in critical cleanroom environments.