ANTIBODY GMP APC MU IGG2B Κ ITCL 100TEST
Catalog # 77780-754
Supplier: Biolegend
New Product
ANTIBODY GMP APC MU IGG2B Κ ITCL 100TEST
Catalog # 77780-754
Supplier: Biolegend
Supplier Number: 260132
New Product
Frequently Bought Together
Cotton Tip Applicators, DUKAL™ Corporation
Supplier: Dukal
Starswabs II® Specimen Collection and Transport Systems
Supplier: Starplex
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Potassium chloride ACS
Supplier: VWR
Corning® Transport Medium
Supplier: Corning
DAPT ≥98% (by NMR)
Supplier: Enzo Life Sciences
VWR® Grade 707 Blotting Paper, Grade 707
Supplier: VWR International
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Supplier: TCI America
Dimethyl sulfoxide, sterile filtered
Supplier: Corning
Accessories for Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
Cellulose, microcrystalline
Supplier: Thermo Scientific Chemicals
Ozone AccuVac® Ampules, MR, Hach
Supplier: Hach
Forskolin ≥99% (by TLC)
Supplier: Enzo Life Sciences
Chloroform ≥99.9% stabilised, OmniSolv®, Supelco®
Supplier: MilliporeSigma