Anti-FANCD2 Rabbit Polyclonal Antibody
Catalog # 77736-614
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:FANCD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9BXW9
- Antigen synonyms:DKFZp762A223|FA 4|FA D2|FA4|FAC D2|FACD 2|FACD|FACD2|FACD2_HUMAN|FAD|FAD2|FANC D2|FANCD 2|FANCD|FANCD2|FANCONI ANEMIA COMPLEMENTATION GROUP D|Fanconi anemia complementation group D2|Fanconi anemia group D2 protein|FANCONI PANCYTOPENIA TYPE 4|FLJ23826|OTTHUMP00000158853|OTTHUMP00000207925|Protein FACD2|Type 4 Fanconi pancytopenia
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:DKFZp762A223
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: FANCD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Frequently Bought Together