Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 $
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MATRIX 1.0ML SCREWTOP TRI-CODE TUBE
Catalog # 77778-528
Supplier:
Thermo Fisher Scientific
New Product
Expand image
MATRIX 1.0ML SCREWTOP TRI-CODE TUBE
Catalog # 77778-528
Supplier:
Thermo Fisher Scientific
Supplier Number:
3741-WP1DPST6
New Product
Frequently Bought Together
epT.I.P.S.® Reloads, Pipette Tips
Supplier: Eppendorf
Optifit Pipette Tips
Supplier: Sartorius
VWR® Gel-Loading Pipette Tips
Supplier: VWR International
Axygen® Research-Grade Pipette Tips, Corning
Supplier: Corning
VWR® Universal Pipette Tips
Supplier: VWR International