ANTIBODY RB X-PTRF A301-270A
Catalog # 77765-208
Supplier: BETHYL LABORATORIES
New Product
ANTIBODY RB X-PTRF A301-270A
Catalog # 77765-208
Supplier: BETHYL LABORATORIES
Supplier Number: A301-270A
New Product
Frequently Bought Together
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
Methanol ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Leakbuster™ Specimen Containers
Supplier: Starplex
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
Water, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Supplier: VWR International
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Glycerin
Supplier: Ward's Science
Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade
Supplier: Thermo Scientific Chemicals
Sodium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Exel International® Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
Glutaraldehyde 25% in aqueous solution, EM Grade
Supplier: Electron Microscopy Sciences
Cetrimide Select Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
HEPES Buffered Saline, Promocell
Supplier: Promocell