ANTIBODY RB X-PRMT3 A302-527A
Catalog # 77766-376
Supplier: BETHYL LABORATORIES
New Product
ANTIBODY RB X-PRMT3 A302-527A
Catalog # 77766-376
Supplier: BETHYL LABORATORIES
Supplier Number: A302-527A
New Product
Frequently Bought Together
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Quality Economy Brand Luer Slip Syringes with Attached Needle, Air-Tite
Supplier: Air-Tite
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Accessories for Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
SYRINGE STERILE WITHOUT NEEDLE 10ML
Supplier: AMX INDUSTRIAL TRADE MX
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
VWR® Dissecting Delicate Scissors
Supplier: VWR International
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA