ANTIBODY RB X-PHC3 A301-570A
Catalog # 77767-046
Supplier: BETHYL LABORATORIES
New Product
ANTIBODY RB X-PHC3 A301-570A
Catalog # 77767-046
Supplier: BETHYL LABORATORIES
Supplier Number: A301-570A
New Product
Frequently Bought Together
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Supplier: MilliporeSigma
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Masterflex® L/S® Precision Pump Tubing, Puri-Flex®, Avantor®
Supplier: VWR International
Petroleum spirit, 35…60 °C, GR ACS, Supelco®
Supplier: MilliporeSigma
Precision pH Test Strips
Supplier: Precision Laboratories
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Difco™ Raka-Ray No.3 Medium, BD
Supplier: BD
Molecular sieve 3A (0.3 nm, 3 Å), pellets Mesh 8 - 12, Ø 1/16", Supelco®
Supplier: MilliporeSigma
Cotton Tip Applicators, DUKAL™ Corporation
Supplier: Dukal
EE Broth Mossel
Supplier: HARDY DIAGNOSTICS CA
Anti-DCX Rabbit Polyclonal Antibody
Supplier: Prosci
Microsep Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Lactose monohydrate for microbiology, Millipore®
Supplier: MilliporeSigma
LabKlenz™ 100 Alkaline Detergents
Supplier: Steris