ANTIBODY RB X-MCEF A302-538A
Catalog # 77773-630
Supplier: BETHYL LABORATORIES
New Product
ANTIBODY RB X-MCEF A302-538A
Catalog # 77773-630
Supplier: BETHYL LABORATORIES
Supplier Number: A302-538A
New Product
Frequently Bought Together
Orthophosphoric acid 25% (v/v) in aqueous solution (1 + 3)
Supplier: Ricca Chemical
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Sodium chloride ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Leakbuster™ Specimen Containers
Supplier: Starplex
LaMotte® InstaTest® Water Test Strips
Supplier: LaMotte
Disodium hydrogen phosphate, anhydrous ≥99.0% ACS
Supplier: VWR
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
Tryptone to 2xYT Broth, Life Technologies
Supplier: BD
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
Lysine Medium, BD Diagnostics
Supplier: BD
ZOMBIE NIR FIXABLE VIABILITY KIT 100 TES
Supplier: Biolegend