ANTIBODY RB X-VPS15 TRIAL A302-570A-T
Catalog # 77768-258
Supplier: BETHYL LABORATORIES
New Product
ANTIBODY RB X-VPS15 TRIAL A302-570A-T
Catalog # 77768-258
Supplier: BETHYL LABORATORIES
Supplier Number: A302-570A-T
New Product
Frequently Bought Together
Counting Chambers, C-Chip, NanoEntek
Supplier: NANOENTEK USA INC.
BD Microtainer® Tubes, BD Diagnostics
Supplier: BD
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Calcium chloride dihydrate 99.0-107.0% USP, Ph. Eur., BP, JP endotoxin tested
Supplier: Avantor Performance Materials
VWR® Disposable Petri Dishes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Potassium hydroxide 0.100 N (N/10) in isopropanol (IPA)
Supplier: Ricca Chemical
Toluene ACS
Supplier: INNOVA CHEMICALS CA
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Corning® DMEM/Ham's F-12 50/50 Mix Products, Corning
Supplier: Corning
Corning® Cell and Tissue Adhesive, Corning
Supplier: Corning
Diversey™ Oxivir® Tb Disinfectant Solution, Antylia Scientific
Supplier: Antyila Scientific
Corning® Trypsin
Supplier: Corning