ANTIBODY RB X-PTPN12/PTP-PEST A301-302AT
Catalog # 77774-960
Supplier: BETHYL LABORATORIES
New Product
ANTIBODY RB X-PTPN12/PTP-PEST A301-302AT
Catalog # 77774-960
Supplier: BETHYL LABORATORIES
Supplier Number: A301-302A-T
New Product
Frequently Bought Together
Acetonitrile ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Sodium sulfate, anhydrous ≥99.0%, white powder ACS, VWR Chemicals BDH®
Supplier: VWR International
Tergazyme® Enzyme-Active Powder Detergents
Supplier: Alconox
VWR® Reagent alcohol
Supplier: VWR International
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Extran® MN 01 Powdered Detergent for Manual Washing, MilliporeSigma
Supplier: MilliporeSigma
Human Recombinant CNTF (from <i>E. coli</i>)
Supplier: Peprotech
Paraformaldehyde 4% in PBS
Supplier: Thermo Scientific Chemicals
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
DIBMA Screening Kit, HEPES
Supplier: CUBE BIOTECH
VWR® Premium Dissecting Scissors with Sharp/Blunt Blades
Supplier: VWR International
Orion™ Sure-Flow® pH Electrode 9172BNWP, Thermo Scientific
Supplier: Thermo Fisher Scientific
Biebrich scarlet
Supplier: Thermo Scientific Chemicals
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Q5® High-Fidelity DNA Polymerase, New England BioLabs
Supplier: New England Biolabs (NEB)
BD Angiocath™ IV Catheters, BD™
Supplier: BD