ANTIBODY RB X-BLM A300-570A
Catalog # 77771-772
Supplier: BETHYL LABORATORIES
New Product
ANTIBODY RB X-BLM A300-570A
Catalog # 77771-772
Supplier: BETHYL LABORATORIES
Supplier Number: A300-570A
New Product
Frequently Bought Together
VWR® Traceable® Conductivity Calibration Standards (CRM)
Supplier: VWR International
Acetonitrile ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Calcium chloride dihydrate 99.0-105.0% ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Axygen® Graduated Transport Tubes, Corning
Supplier: Corning
Single-Use Needles, BD Medical
Supplier: BD
VWR® Reagent alcohol
Supplier: VWR International
Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Supplier: Corning
Glycerin
Supplier: Ward's Science
Sulfuric acid standardised solution 1.0 N in aqueous solution
Supplier: Thermo Scientific Chemicals
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Lactobacilli MRS Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
Dimethyl sulfoxide ≥99.9% ACS, VWR Chemicals BDH®
Supplier: VWR International