CAPTURE BEAD LPX HU OPG A4 13X 70UL
Catalog # CA77759-760
Supplier: Biolegend
New Product
CAPTURE BEAD LPX HU OPG A4 13X 70UL
Catalog # CA77759-760
Supplier: Biolegend
Supplier Number: 741368
New Product
Frequently Bought Together
Alconox® Powdered Precision Cleaners
Supplier: Alconox
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
Methanol, anhydrous ≥99.8% (by GC), GR ACS ACS, Supelco®
Supplier: MilliporeSigma
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Cytology Funnels for Shandon CytoSpin™ Centrifuge, Biomedical Polymers
Supplier: Biomedical Polymers
LaMotte® InstaTest® Water Test Strips
Supplier: LaMotte
Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Supplier: Corning
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Reagent alcohol
Supplier: VWR International
Tryptone to 2xYT Broth, Life Technologies
Supplier: BD
Sodium dodecyl sulfate (SDS) ≥99.0%, OmniPur®, Millipore®
Supplier: MilliporeSigma