CAPTURE BEAD NHP IFN-A2 B3 13X 70UL
Catalog # CA77759-842
Supplier: Biolegend
New Product
CAPTURE BEAD NHP IFN-A2 B3 13X 70UL
Catalog # CA77759-842
Supplier: Biolegend
Supplier Number: 741425
New Product
Frequently Bought Together
Leakbuster™ Specimen Containers
Supplier: Starplex
Alcotabs® Critical Cleaning Detergent Tablets
Supplier: Alconox
Sodium chloride ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Masterflex® L/S® Precision Pump Tubing, Versilon™ 2001, Avantor®
Supplier: VWR International
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
ZOMBIE AQUA FIXABLE VIABILITY KIT 100 TE
Supplier: Biolegend
Pharmacy Bags, Reclosable, Elkay Plastics
Supplier: Elkay Plastics
Magnesium Chloride Hexahydrate
Supplier: Ward's Science
ACETIC RINSE SOLUTION 4L
Supplier: Ostrem Chemical
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Slide Tray Racks and Aluminum Slide Trays, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Micrewtube® Microcentrifuge Tube with Screw Cap, Simport Scientific
Supplier: Simport Scientific
Lance Blades, Cincinnati Surgical
Supplier: CINCINNATI SURGICAL CO.