CAPTURE BEAD CXCL8 IL-8 A4 13X 70UL
Catalog # CA77759-280
Supplier: Biolegend
New Product
CAPTURE BEAD CXCL8 IL-8 A4 13X 70UL
Catalog # CA77759-280
Supplier: Biolegend
Supplier Number: 740988
New Product
Frequently Bought Together
Water, deionized VWR®
Supplier: VWR
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Leakbuster™ Specimen Containers
Supplier: Starplex
Easy Bleach™ Disinfectant Tablets, Berkshire
Supplier: Berkshire
Polyethylene Disposable Apron
Supplier: Dival Safety & Supplies
Lecithin, refined
Supplier: Thermo Scientific Chemicals
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
SpecCheck Secondary Gel Standards Sets, DPD Chlorine, LR
Supplier: Hach
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
SYRINGE WITH NEEDLE 5ML
Supplier: AMX INDUSTRIAL TRADE MX
LabKlenz™ 250 Acid Detergents
Supplier: Steris
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
INDICATOR PAPER WATOR DARK BLUE 78X40 MM
Supplier: Macherey-Nagel
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Supplier: TCI America
CL17 DPD Indicator, Hach
Supplier: Hach