LPX RAT GM-CSF CAPTURE BEAD A10 13X 70UL
Catalog # CA77759-634
Supplier: Biolegend
New Product
LPX RAT GM-CSF CAPTURE BEAD A10 13X 70UL
Catalog # CA77759-634
Supplier: Biolegend
Supplier Number: 741267
New Product
Frequently Bought Together
Extracta™ DNA Prep for PCR, Quantabio
Supplier: Quantabio
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Blood Bank Saline Solutions
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Mounting Bracket For Qlicksmart® BladeFLASK Blade Remover, Mortech
Supplier: Mortech Manufacturing
Toluene ACS
Supplier: INNOVA CHEMICALS CA
Accessories for Micro-Osmometer, Advanced Instruments
Supplier: Advanced Instruments
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Corning® Matrigel® Basement Membrane Matrix, Corning
Supplier: Corning
Difco™ Agar Soldifying Agent, BD Diagnostics
Supplier: BD