LPLEX HU CXCL10 (IP-10) BEAD B3 13X 70UL
Catalog # CA77759-808
Supplier: Biolegend
New Product
LPLEX HU CXCL10 (IP-10) BEAD B3 13X 70UL
Catalog # CA77759-808
Supplier: Biolegend
Supplier Number: 741408
New Product
Frequently Bought Together
BD GasPak™ EZ Container Systems, BD Diagnostics
Supplier: BD
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Triethylamine ≥99.5% for HPLC, Supelco®
Supplier: MilliporeSigma
Vesphene® III st Disinfectant Cleaners
Supplier: Steris
Orion™ pH Buffer Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
First Aid Central Federal Regulation First Aid Kits, Acme United
Supplier: ACME UNITED LIMITED CA
Miltex® BiopBlade, Integra
Supplier: Miltex
Chloroform ≥99.9% stabilised, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Sodium dodecyl sulfate (SDS) ≥99.0%, OmniPur®, Millipore®
Supplier: MilliporeSigma
PerfeCTa® qPCR ToughMix®, QuantaBio
Supplier: Quantabio
LabKlenz™ 250 Acid Detergents
Supplier: Steris
Carbon disulfide ≥99.9%, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Sandwich Bags, Low Density Flip Top Elkay Plastics
Supplier: Elkay Plastics
1-Octanesulfonic acid sodium salt, Supelco®
Supplier: MilliporeSigma
Dithiothreitol (DTT, Cleland's reagent) ≥98% (by iodometry)
Supplier: Enzo Life Sciences