CAPTURE BEAD HU IL-1B B613X 70UL
Catalog # CA77759-224
Supplier: Biolegend
New Product
CAPTURE BEAD HU IL-1B B613X 70UL
Catalog # CA77759-224
Supplier: Biolegend
Supplier Number: 740669
New Product
Frequently Bought Together
Water, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Cotton Tip Applicators, DUKAL™ Corporation
Supplier: Dukal
Sterile IPA 70% Spray, ITW Texwipe
Supplier: ITW Texwipe
Single-Use Needles, BD Medical
Supplier: BD
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Quality Economy Brand Luer Lock Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Corning® Matrigel® Basement Membrane Matrix, Corning
Supplier: Corning